missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethane, 99.8%, for HPLC, stabilized with amylene, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 268330010

 View more versions of this product

Catalog No. AC268330010



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For HPLC
Specifications

Specifications

Dichloromethane
(1 cm cell vs HPLC−grade water)
(1 cm cell vs HPLC-grade water)
99.8%
CH2Cl2
01, 60
15, 6135
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
C(Cl)Cl
84.927
CHEBI:15767
Liquid
HPLC
Yes
Glass bottle
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
about 100ppm amylene
0.43 mPa.s (20°C)
Undesignated
1L
Stabilized
75-09-2,513-35-9
0.0001 meq/g max.
CH2Cl2
MFCD00000881
01,676; 02,273; 04,337; 07,239; 13,106
aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
6344
84.93
99.8%
1.3250g/mL
0.0005% max.
0.0002% max.
1.325
0.02% max.
39.0°C to 40.0°C
-97.0°C

Chemical Identifiers

75-09-2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
dichloromethane
CH2Cl2
MFCD00000881
aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine
CHEBI:15767
C(Cl)Cl
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.

GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

Warning

EINECSNumber : 200-838-9

RTECSNumber : PA8050000

TSCA : TSCA

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only