Learn More
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.8%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine, 1L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless, >120 deg.C
Supplier: Thermo Scientific Chemicals 348460010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dichloromethane | |
>120°C | |
Colorless | |
39.0°C to 40.0°C | |
99.8% | |
CH2Cl2 | |
MFCD00000881 | |
01, 60 | |
15, 6135 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
ClCCl | |
84.93 | |
6344 | |
84.93 | |
99.8% | |
Liquid |
75-09-2,513-35-9 | |
-97.0°C | |
1.3250g/mL | |
Authentic | |
AcroSeal™ Glass Bottle | |
CH2Cl2 | |
1 L | |
01,676; 02,273; 04,337; 07,239; 13,106 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
50ppm (approx.) amylene | |
CHEBI:15767 | |
475hPa at 20°C | |
Extra Dry over Molecular Sieve |
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.
GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.
Warning
RUO – Research Use Only