missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethane, 99.8+%, for analysis, stabilized with amylene, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC113460050

 View more versions of this product

Catalog No. AC113460050



Description

Description

For analysis
Specifications

Specifications

Dichloromethane
0.0003 meq/g max.
CH2Cl2
MFCD00000881
01,676; 02,273; 04,337; 07,239; 13,106
aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
6344
84.93
≥99.8;%
0.05ppm max.
0.3ppm max.
0.1ppm max.
Plastic drum
1.325
5 APHA max.
0.1ppm max.
39.0°C to 40.0°C
-97.0°C
75-09-2,513-35-9
99.8+%
CH2Cl2
01, 60
15, 6135
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
C(Cl)Cl
84.927
CHEBI:15767
Liquid
Analysis
1.3250g/mL
0.1ppm max.
0.1ppm max.
0.001% max.
15 to 50ppm amylene
0.02% max. (Coulometric)
0.43 mPa.s (20°C)
Undesignated
5L

Chemical Identifiers

75-09-2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
dichloromethane
CH2Cl2
MFCD00000881
aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine
CHEBI:15767
C(Cl)Cl
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.

GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

Warning

SDS Documents