Please login to your online account to display your discounted pricing

Dichloromethane, >99.8%, Alfa Aesar™

Suitable For Pesticide Residue Analysis

$57.27 - $57.27

Chemical Identifiers

Chemical Name or Material Dichloromethane
CAS 75-09-2
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range >99.8%
Molecular Formula CH2Cl2
Formula Weight 84.93
MDL Number MFCD00000881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42006K2

 
alfa aesar
42006K2
1L Each for $57.27

AA42006K7

 
alfa aesar
42006K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42006M1

 
alfa aesar
42006M1
4 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42006M6

 
alfa aesar
42006M6
4 x 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 84.93
Color Clear colorless
Quantity 1L
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Dichloromethane
EINECS Number 200-838-9
Ignition Point 605°C (1121°F)
Purity Grade Environmental Grade
Assay Percent Range >99.8%
Molecular Formula CH2Cl2
Odor Sweet
MDL Number MFCD00000881
CAS 75-09-2
Vapor Pressure 475 hPa (356mm Hg) at 20°C
Health Hazard 1 H351: Suspected of causing cancer.
Appearance Colorless liquid
Density 1.325g/cm3 at 20°C

Maximum level of impurities: Color (APHA) 10, Cl 10ppm, Halogenated residue 10ng/L heptachlorepoxide, Evaporation residue 3ppm, H2O 0.02%

Safety and Handling

H351: Suspected of causing cancer.