missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethane, 99.5% min., ACS, EMD Millipore

$95.40 - $5,082.43

Chemical Identifiers

CAS 75-09-2
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula CH2Cl2
Formula Weight 84.93
Synonym Methylene Chloride
Chemical Name or Material Dichloromethane, For ACS analysis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MDX08356 Emd Millipore Corporation
DX08356
500mL Each for $95.40
MDX08353 Emd Millipore Corporation
DX0835-3
4L Each for $254.74
MDX083520 Emd Millipore Corporation
DX083520
200L Each for $5,082.43
MDX08355 Emd Millipore Corporation
DX08355
20L Each for $710.60
Specifications Description & Specifications

Chemical Identifiers

75-09-2
99.5% min.
84.93
Dichloromethane, For ACS analysis
Liquid
CH2Cl2
Methylene Chloride

Specifications

75-09-2
99.5% min.
84.93
Clear colorless
Dichloromethane, For ACS analysis
Liquid
CH2Cl2
Methylene Chloride
500mL