Learn More
Dichloromalononitrile 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113450050
Specifications
Dichloromalononitrile | |
Authentic | |
1.4310 to 1.4360 (20°C, 589nm) | |
5g | |
13063-43-9 | |
C3Cl2N2 | |
MFCD00001841 | |
2,2-dichloromalononitrile, dichloro-malononitril, dichlorodicyanomethane, dichloromalonitrile, dichloromalononitrile, dichloropropanedinitrile, malononitrile, dichloro, propanedinitrile, dichloro, propanedinitrile,2,2-dichloro, wln: ncxggcn | |
C(#N)C(C#N)(Cl)Cl | |
134.947 | |
134.95 | |
≥96.0% |
1.3100g/mL | |
Glass Bottle | |
Yellow to Brown | |
97% | |
97% | |
NCCCl2CN | |
02, II, 537 | |
NCWGPYPDPWOJKC-UHFFFAOYSA-N | |
2,2-dichloropropanedinitrile | |
25660 | |
Liquid |
Chemical Identifiers
13063-43-9 | |
134.947 | |
NCWGPYPDPWOJKC-UHFFFAOYSA-N | |
25660 | |
C(#N)C(C#N)(Cl)Cl |
C3Cl2N2 | |
MFCD00001841 | |
2,2-dichloromalononitrile, dichloro-malononitril, dichlorodicyanomethane, dichloromalonitrile, dichloromalononitrile, dichloropropanedinitrile, malononitrile, dichloro, propanedinitrile, dichloro, propanedinitrile,2,2-dichloro, wln: ncxggcn | |
2,2-dichloropropanedinitrile |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 235-955-4
RTECSNumber : OO3740000