Please login to your online account to display your discounted pricing

Dichloromalononitrile 97%, ACROS Organics™

$168.33 - $168.33

Chemical Identifiers

Chemical Name or Material Dichloromalononitrile
Name Note 97%
CAS 13063-43-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3Cl2N2
Linear Formula NCCCl2CN
Formula Weight 134.95
MDL Number MFCD00001841
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113450050

  SDS
thermo fisher scientific
113450050
Glass bottle 5g Each for $168.33
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 134.95
Quantity 5g
Color Clear yellow to light brown
Beilstein 02, II, 537
RTECS Number OO3740000
EINECS Number 235-955-4
Density 1.3100g/mL
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.4310 to 1.4360 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Chemical Name or Material Dichloromalononitrile
CAS 13063-43-9
Linear Formula NCCCl2CN
MDL Number MFCD00001841
Name Note 97%
Molecular Formula C3Cl2N2
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.