Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloromalononitrile, 96%

Chemical Identifiers

Chemical Name or Material Dichloromalononitrile
CAS 13063-43-9
Molecular Formula C3Cl2N2
Formula Weight 134.95
MDL Number MFCD00001841
UN Number UN3276
Beilstein 906848
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2177903

  SDS
alfa aesar™
B2177903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2177906

  SDS
alfa aesar™
B2177906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2177914

  SDS
alfa aesar™
B2177914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.95
Chemical Name or Material Dichloromalononitrile
Molecular Formula C3Cl2N2
Odor Pungent
MDL Number MFCD00001841
CAS 13063-43-9
Beilstein 906848
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
RTECS Number OO3740000
Boiling Point 96° to 97°C
Assay 96%
UN Number UN3276
Health Hazard 2 GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

2° to 8°C