Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloroisobutylmethylsilane, 97%

Chemical Identifiers

Chemical Name or Material Dichloroisobutylmethylsilane
CAS 18028-96-1
Molecular Formula C5H12Cl2Si
Formula Weight 171.14
MDL Number MFCD00083134
UN Number UN2987
Synonym Isobutylmethyldichlorosilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1653306

 
alfa aesar™
L1653306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1653314

 
alfa aesar™
L1653314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1653322

 
alfa aesar™
L1653322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.14
Chemical Name or Material Dichloroisobutylmethylsilane
Assay 97%
Molecular Formula C5H12Cl2Si
CAS 18028-96-1
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
MDL Number MFCD00083134
UN Number UN2987
Synonym Isobutylmethyldichlorosilane
Boiling Point 130°C
Quantity 5g
Flash Point 64°C (147°F)
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.