Please login to your online account to display your discounted pricing

Dichlorofluorescein TS, 0.1% (w/v) in Dilute Alcohol, Ricca Chemical

$86.24 - $161.06

Chemical Identifiers

Chemical Name or Material Dichlorofluorescein
Name Note Test Solution
CAS 64-17-5
CAS Min % 53.0
CAS Max % 56.0
CAS 7732-18-5
CAS Min % 44.48
CAS Max % 46.3
CAS 76-54-0
CAS Min % 0.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2549-16

 
ricca chemical company
RI254916
Poly natural 500mL Each for $161.06

2549-4

 
ricca chemical company
RI25494
Poly natural 120mL Each for $86.24
Description & Specifications

Specifications

Physical Form Liquid
Color Flourescent orange-yellow
Quantity 500mL
Concentration or Composition Notes 0.1% (w/v)
Solvent or Matrix Dilute Alcohol
Chemical Name or Material Dichlorofluorescein
CAS Max % 0.12
CAS 1310-73-2
CAS Min % 0.02
CAS Max % 0.02
CAS 7732-18-5 (Water, Deionized)
Name Note Test Solution
CAS 64-17-5
CAS Min % 53.0
Packaging Poly natural
CAS Max % 56.0
CAS Min % 44.48
CAS Max % 46.3
CAS 76-54-0
CAS Min % 0.11
CAS 7732-18-5