Please login to your online account to display your discounted pricing

Dichlorofluorescein, ACS Grade, LabChem™

$219.20 - $219.20

Chemical Identifiers

Chemical Name or Material Dichlorofluorescein
CAS 76-54-0
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C20H10Cl2O5
Formula Weight 401.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC135707

 
lab chem
LC135707
Amber Glass 10g Each for $219.20
Description & Specifications

Specifications

Formula Weight 401.2
Health Hazard 1 Warning
CAS 76-54-0
Purity Grade ACS Grade
Density 790kg/m3
EINECS Number 200-968-6
Solubility Soluble in water
Color Orange to red-brown
Recommended Storage Room Temperature
Chemical Name or Material Dichlorofluorescein
Melting Point 280°C
CAS Max % 100
Identification Passes Test
Decomposition Information Carbon dioxide; Carbon monoxide; Chlorine
Specific Gravity 790kg/m3
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Suitability Passes Test
Packaging Amber Glass
Physical Form Powder
Linear Formula C20H10Cl2O5
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 10g
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature