missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloroethylmethylsilane, 94%

$60.15 - $217.43

Chemical Identifiers

CAS 4525-44-4
Molecular Formula C3H8Cl2Si
Molecular Weight (g/mol) 143.082
MDL Number MFCD00013607
InChI Key PNECSTWRDNQOLT-UHFFFAOYSA-N
Synonym ethylmethyldichlorosilane, methylethyldichlorosilane, dichloro ethyl methylsilane, silane, dichloroethylmethyl, dichloro ethyl methyl silane, unii-47rob570jw, acmc-20akvy, dichloro-ethyl-methyl-silane, dichloro ethyl ?methylsilane, dichloro-ethyl-methyl silane
PubChem CID 78286
IUPAC Name dichloro-ethyl-methylsilane
SMILES CC[Si](C)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1649309 View Documents Alfa Aesar™
L1649309
10g Each for $60.15
 
AAL1649318 View Documents Alfa Aesar™
L1649318
50g Each for $217.43
 
Specifications

Chemical Identifiers

4525-44-4
143.082
PNECSTWRDNQOLT-UHFFFAOYSA-N
78286
CC[Si](C)(Cl)Cl
C3H8Cl2Si
MFCD00013607
ethylmethyldichlorosilane, methylethyldichlorosilane, dichloro ethyl methylsilane, silane, dichloroethylmethyl, dichloro ethyl methyl silane, unii-47rob570jw, acmc-20akvy, dichloro-ethyl-methyl-silane, dichloro ethyl ?methylsilane, dichloro-ethyl-methyl silane
dichloro-ethyl-methylsilane

Specifications

4525-44-4
MFCD00013607
1361374
PNECSTWRDNQOLT-UHFFFAOYSA-N
dichloro-ethyl-methylsilane
78286
94%
100°C
2°C (35°F)
C3H8Cl2Si
UN2985
ethylmethyldichlorosilane, methylethyldichlorosilane, dichloro ethyl methylsilane, silane, dichloroethylmethyl, dichloro ethyl methyl silane, unii-47rob570jw, acmc-20akvy, dichloro-ethyl-methyl-silane, dichloro ethyl ?methylsilane, dichloro-ethyl-methyl silane
CC[Si](C)(Cl)Cl
143.082
143.09
Acrid
10g
Dichloroethylmethylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA