Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloroethylenebis(indenyl)zirconium(IV), racemic mixture

Useful for the preparation of metallocene type catalysts

Chemical Identifiers

Chemical Name or Material Dichloroethylenebis(indenyl)zirconium(IV)
CAS 100080-82-8
Name Note Racemic mixture
Formula Weight 418.48
MDL Number MFCD01314544
UN Number UN3261
Synonym Ethylenebis(indenyl)zirconium(IV) dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4044902

  SDS
alfa aesar™
4044902
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4044927

  SDS
alfa aesar™
4044927
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 418.48
CAS 100080-82-8
Chemical Name or Material Dichloroethylenebis(indenyl)zirconium(IV)
MDL Number MFCD01314544
UN Number UN3261
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture Sensitive
Health Hazard 1 Danger
Name Note Racemic mixture
Physical Form Powder
Synonym Ethylenebis(indenyl)zirconium(IV) dichloride
Quantity 0.5g
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.