missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorodiphenylsilane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113341000

 View more versions of this product

Catalog No. AC113341000



Description

Description

Specifications

Specifications

Dichlorodiphenylsilane
155°C
Glass bottle
1.204
Brown or Colorless to Yellow
100mL
96% min. (GC)
(C6H5)2SiCl2
16,91
benzene, 1,1'-dichlorosilylene bis, dichlor-difenylsilan, dichlor-difenylsilan czech, dichloro diphenyl silane, diphenyl dichlorosilane, diphenyldichlorosilane, diphenylsilicon dichloride, diphenylsilyl dichloride, hsdb 316, silane, dichlorodiphenyl
OSXYHAQZDCICNX-UHFFFAOYSA-N
dichloro(diphenyl)silane
6627
Liquid
1.2040g/mL
Authentic
1.5765 to 1.5815
305.0°C
-22.0°C
80-10-4
C12H10Cl2Si
MFCD00000489
13,74
Solubility in water: decomposes
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
253.20
253.2
97%

Chemical Identifiers

80-10-4
253.20
OSXYHAQZDCICNX-UHFFFAOYSA-N
6627
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
C12H10Cl2Si
MFCD00000489
benzene, 1,1'-dichlorosilylene bis, dichlor-difenylsilan, dichlor-difenylsilan czech, dichloro diphenyl silane, diphenyl dichlorosilane, diphenyldichlorosilane, diphenylsilicon dichloride, diphenylsilyl dichloride, hsdb 316, silane, dichlorodiphenyl
dichloro(diphenyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

EINECSNumber : 201-251-

SDS Documents
Product Certifications