missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorodiphenylmethane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC113330100

 View more versions of this product

Catalog No. AC113330100



Description

Description

Specifications

Specifications

Dichlorodiphenylmethane
>112°C
Glass bottle
1.23
Yellow
97%
96% min. (GC)
(C6H5)2CCl2
05, 590
Solubility in water: reacts. Other solubilities: soluble in ether,benzene,ethanol and toluene
C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl
237.13
237.13
97%
1.2300g/mL
Authentic
1.6030 to 1.6050
305.0°C
10mL
2051-90-3
C13H10Cl2
MFCD00000811
alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, benzophenone dichloride, dichloro phenyl methyl benzene, dichlorodiphenyl methane, dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, methane, dichlorodiphenyl, unii-81fez29ak6
OPTDDWCXQQYKGU-UHFFFAOYSA-N
[dichloro(phenyl)methyl]benzene
16327
Liquid

Chemical Identifiers

2051-90-3
237.13
OPTDDWCXQQYKGU-UHFFFAOYSA-N
16327
C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl
C13H10Cl2
MFCD00000811
alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, benzophenone dichloride, dichloro phenyl methyl benzene, dichlorodiphenyl methane, dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, methane, dichlorodiphenyl, unii-81fez29ak6
[dichloro(phenyl)methyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 218-134-5

TSCA : TSCA

SDS Documents
Product Certifications