Learn More
Dichlorodiphenylmethane, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113332500
Specifications
Dichlorodiphenylmethane | |
>112°C | |
Glass bottle | |
1.23 | |
Yellow | |
97% | |
96% min. (GC) | |
(C6H5)2CCl2 | |
05, 590 | |
Solubility in water: reacts. Other solubilities: soluble in ether,benzene,ethanol and toluene | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl | |
237.13 | |
237.13 | |
97% |
1.2300g/mL | |
Authentic | |
1.6030 to 1.6050 | |
305.0°C | |
250mL | |
2051-90-3 | |
C13H10Cl2 | |
MFCD00000811 | |
alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, benzophenone dichloride, dichloro phenyl methyl benzene, dichlorodiphenyl methane, dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, methane, dichlorodiphenyl, unii-81fez29ak6 | |
OPTDDWCXQQYKGU-UHFFFAOYSA-N | |
[dichloro(phenyl)methyl]benzene | |
16327 | |
Liquid |
Chemical Identifiers
2051-90-3 | |
237.13 | |
OPTDDWCXQQYKGU-UHFFFAOYSA-N | |
16327 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl |
C13H10Cl2 | |
MFCD00000811 | |
alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, benzophenone dichloride, dichloro phenyl methyl benzene, dichlorodiphenyl methane, dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, methane, dichlorodiphenyl, unii-81fez29ak6 | |
[dichloro(phenyl)methyl]benzene |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 218-134-5
TSCA : TSCA