missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorodimethylsilane, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC113310025

 View more versions of this product

Catalog No. AC113310025



Description

Description

Specifications

Specifications

Dichlorodimethylsilane
−9°C
Glass bottle
1.06
70.0°C
-76.0°C
75-78-5
C2H6Cl2Si
03,114; 04,183; 17,113
Solubility in water: reacts. Other solubilities: soluble in proton-free solvents (care must be,taken with ketones and esters),soluble in benzene and ether,decomposes in ethanol
C[Si](C)(Cl)Cl
129.06
129.06
99+%
1.0600g/mL
Authentic
1.4030 to 1.4050
0.6 mPa.s (25°C)
Colorless to Yellow
2.5L
99% min. (GC)
(CH3)2SiCl2
dichloro dimethyl silane, dichlorodimethylsilicon, dimethyl dichlorosilane, dimethyl-dichlorsilan, dimethyldichlorosilane, dimethylsilane dichloride, inerton aw-dmcs, inerton dw-dmc, repel-silan, silane, dichlorodimethyl
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dichloro(dimethyl)silane
6398
Liquid

Chemical Identifiers

75-78-5
129.06
dichloro dimethyl silane, dichlorodimethylsilicon, dimethyl dichlorosilane, dimethyl-dichlorsilan, dimethyldichlorosilane, dimethylsilane dichloride, inerton aw-dmcs, inerton dw-dmc, repel-silan, silane, dichlorodimethyl
dichloro(dimethyl)silane
C2H6Cl2Si
LIKFHECYJZWXFJ-UHFFFAOYSA-N
6398
C[Si](C)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 200-901-

SDS Documents