Please login to your online account to display your discounted pricing

Dichlorodimethylgermane, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC316500050

 View more versions of this product

Catalog No. AC316500050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Quantity 5g
Physical Form Liquid
Formula Weight 173.57
Color Colorless
Density 1.5000g/mL
UN Number 2920
Linear Formula (CH3)2GeCl2
Synonym Dimethylgermanium dichloride
Molecular Formula C2H6Cl2Ge
Assay Percent Range 97%
Chemical Name or Material Dichlorodimethylgermane
CAS 1529-48-2
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00013588
Packaging Glass bottle
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dichlorodimethylgermane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2H6Cl2Ge
Linear Formula (CH3)2GeCl2
Formula Weight 173.57
MDL Number MFCD00013588
Synonym Dimethylgermanium dichloride
Accessories

GHS Signal Word: Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents