missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichlorobis(triphenylphosphine)nickel(II), 98%

Manufacturer:  Alfa Aesar™ 1393009

 View more versions of this product

Catalog No. AA1393009



Description

Description

  • Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron
Specifications

Specifications

Dichlorobis(triphenylphosphine)nickel(II)
Moisture Sensitive
~225°C (decomposition)
14264-16-5
MFCD00009592
ZBRJXVVKPBZPAN-UHFFFAOYSA-N
nickel(2+);triphenylphosphane;dihydrochloride
131664342
Crystalline Powder
Odorless
Black to Gray
10g
C36H32Cl2NiP2+2
bis triphenylphosphine dichloronickel ii
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl.Cl.[Ni+2]
656.193
654.18
98%

Chemical Identifiers

14264-16-5
656.193
ZBRJXVVKPBZPAN-UHFFFAOYSA-N
131664342
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl.Cl.[Ni+2]
C36H32Cl2NiP2+2
MFCD00009592
bis triphenylphosphine dichloronickel ii
nickel(2+);triphenylphosphane;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : QR6170000

TSCA : TSCA

Documents