Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichlorobis(tricyclohexylphosphine)palladium(II), Pd 14.4%

$104.42 - $104.42

Chemical Identifiers

Chemical Name or Material Dichlorobis (tricyclohexylphosphine)palladium (II)
CAS 29934-17-6
Molecular Formula PdCl2[P(C6H12)3]2
Formula Weight 744.24
MDL Number MFCD00191830
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4484403

  SDS
alfa aesar™
4484403
1g Each for $104.42
Description & Specifications

Specifications

Chemical Name or Material Dichlorobis (tricyclohexylphosphine)palladium (II)
Concentration Pd 14.4%
Quantity 1g
CAS 29934-17-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
Molecular Formula PdCl2[P(C6H12)3]2
Formula Weight 744.24
MDL Number MFCD00191830
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.