Learn More
Dichlorobis(tri-o-tolylphosphine)palladium(II), 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04497603
Description
Dichlorobis(tri-o-tolylphosphine)palladium(II) is used in reaction of tributyltin enolates, prepared in situ from tributyltin methoxide and enol acetates, with aryl bromides, coupling reaction of aryl bromides with vinylic acetates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dichlorobis (tri-o-tolylphosphine)palladium (II) | |
Yellow | |
MFCD00274659 | |
Air and moisture sensitive | |
Soluble in ethanol,benzene,and ethereal solvents. | |
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1 | |
786.06 | |
786.08 | |
Powder |
40691-33-6 | |
C42H42Cl2P2Pd | |
1 g | |
palladium,dichlorobis tris 2-methylphenyl phosphine | |
GMLMZLGOMMIIMA-UHFFFAOYSA-L | |
palladium;tris(2-methylphenyl)phosphane;dichloride | |
90656943 | |
98% |
Chemical Identifiers
40691-33-6 | |
786.06 | |
GMLMZLGOMMIIMA-UHFFFAOYSA-L | |
90656943 | |
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1 |
C42H42Cl2P2Pd | |
MFCD00274659 | |
palladium,dichlorobis tris 2-methylphenyl phosphine | |
palladium;tris(2-methylphenyl)phosphane;dichloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only