missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichlorobis(tri-o-tolylphosphine)palladium(II), 98%

Manufacturer:  Alfa Aesar™ 4497606

 View more versions of this product

Catalog No. AA4497606



Specifications

Specifications

Dichlorobis (tri-o-tolylphosphine)palladium (II)
Yellow
40691-33-6
MFCD00274659
KFQYTZDYGOBBOE-UHFFFAOYSA-L
palladium;tris(2-methylphenyl)phosphane;dichloride
90656943
Powder
Air & Moisture Sensitive
5g
C42H42Cl2P2Pd-2
palladium,dichlorobis tris 2-methylphenyl phosphine
CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.[Cl-].[Cl-].[Pd]
786.066
786.08
98%

Chemical Identifiers

40691-33-6
786.066
KFQYTZDYGOBBOE-UHFFFAOYSA-L
90656943
CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.[Cl-].[Cl-].[Pd]
C42H42Cl2P2Pd-2
MFCD00274659
palladium,dichlorobis tris 2-methylphenyl phosphine
palladium;tris(2-methylphenyl)phosphane;dichloride
SDS Documents