Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichlorobis(pyridine)palladium(II), 98%

$206.43 - $935.70

Chemical Identifiers

Chemical Name or Material Dichlorobis(pyridine)palladium(II)
CAS 14872-20-9
Molecular Formula C10H10Cl2N2Pd
Formula Weight 335.53
MDL Number MFCD00792248
Synonym Bis(pyridine)palladium(II) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2066303

  SDS
alfa aesar™
B2066303
1g Each for $206.43

AAB2066306

  SDS
alfa aesar™
B2066306
5g Each for $935.70
Description & Specifications

Specifications

Formula Weight 335.53
Chemical Name or Material Dichlorobis(pyridine)palladium(II)
Health Hazard 1 Danger
Molecular Formula C10H10Cl2N2Pd
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
MDL Number MFCD00792248
CAS 14872-20-9
Assay 98%
Synonym Bis(pyridine)palladium(II) chloride
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.