missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloroacetyl chloride, 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 113085000

 View more versions of this product

Catalog No. AC113085000



Specifications

Specifications

Dichloroacetyl chloride
66°C
Glass bottle
1.53
Colorless to Yellow
97%
96% min. (GC)
Cl2CHCOCl
02, 204
acetyl chloride, dichloro
FBCCMZVIWNDFMO-UHFFFAOYSA-N
2,2-dichloroacetyl chloride
6593
147.39
97%
1.5300g/mL
Authentic
1.4600 to 1.4620
107.0°C to 108.0°C
500mL
79-36-7
C2HCl3O
MFCD00000840
15, 3062
Solubility in water: may decompose. Other solubilities: soluble in ether,decomposes in alcohol
C(C(=O)Cl)(Cl)Cl
147.39
CHEBI:34688
Liquid

Chemical Identifiers

79-36-7
147.39
FBCCMZVIWNDFMO-UHFFFAOYSA-N
6593
2,2-dichloroacetyl chloride
C2HCl3O
MFCD00000840
acetyl chloride, dichloro
CHEBI:34688
C(C(=O)Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-199-9

RTECSNumber : AO6650000

TSCA : TSCA

SDS Documents