Learn More
Dichloroacetonitrile, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1061206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dichloroacetonitrile | |
1.369 | |
35°C (95°F) | |
C2HCl2N | |
Cl2CHCN | |
5 g | |
1739029 | |
STZZWJCGRKXEFF-UHFFFAOYSA-N | |
2,2-dichloroacetonitrile | |
18177 | |
109.94 |
3018-12-0 | |
112°C to 114°C | |
Pungent | |
1.4405 | |
MFCD00040886 | |
UN2920 | |
dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562 | |
C(#N)C(Cl)Cl | |
109.937 | |
CHEBI:82444 | |
≥98% |
Chemical Identifiers
3018-12-0 | |
109.937 | |
STZZWJCGRKXEFF-UHFFFAOYSA-N | |
18177 | |
2,2-dichloroacetonitrile |
C2HCl2N | |
MFCD00040886 | |
dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562 | |
CHEBI:82444 | |
C(#N)C(Cl)Cl |
Safety and Handling
GHS H Statement
H314-H318-H226-H341-H361-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 221-159-4
RTECSNumber : AL8465000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only