Please login to your online account to display your discounted pricing

Dichloroacetic anhydride, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  165120500

 View more versions of this product

Catalog No. AC165120500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid After Melting
Formula Weight 239.87
Quantity 50g
Color Clear colorless
Beilstein 02, 204
TSCA TSCA
RTECS Number AG6300000
EINECS Number 223-924-8
Density 1.57g/mL
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Refractive Index 1.4815 to 1.4845 (20°C, 589nm)
CAS 4124-30-5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Dichloroacetic anhydride
Molecular Formula C4H2Cl4O3
Assay Percent Range 98%
Linear Formula (Cl2CHCO)2O
Name Note 98%
MDL Number MFCD00000842

Specifications

Chemical Name or Material Dichloroacetic anhydride
Name Note 98%
CAS 4124-30-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H2Cl4O3
Linear Formula (Cl2CHCO)2O
Formula Weight 239.87
MDL Number MFCD00000842
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

SDS & Safety Documents