missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloroacetic acid, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1474022

 View more versions of this product

Catalog No. AAA1474022



Description

Description

Solubility
Miscible with water, ethanol, diethyl ether, n-octanol and acetone.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Specifications

Dichloroacetic acid
Pungent
9°C to 11°C
79-43-6
C2H2Cl2O2
UN1764
14,3050
JXTHNDFMNIQAHM-UHFFFAOYSA-N
2,2-dichloroacetic acid
6597
128.94
99%
>110°C (230°F)
192°C to 194°C
100g
99%
MFCD00004223
1098596
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
C(C(=O)O)(Cl)Cl
128.936
CHEBI:36386
Liquid

Chemical Identifiers

79-43-6
128.936
JXTHNDFMNIQAHM-UHFFFAOYSA-N
6597
2,2-dichloroacetic acid
C2H2Cl2O2
MFCD00004223
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
CHEBI:36386
C(C(=O)O)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETIC ACID

RTECSNumber : AG6125000

TSCA : TSCA

storageNote1 : Hygroscopic

SDS Documents