missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dichloroacetic acid, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC113060025
Specifications
Dichloroacetic acid | |
>112°C | |
Glass bottle | |
1.56 | |
Brown | |
2.5L | |
99+% | |
Cl2CHCO2H | |
02,202 | |
15,3059 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol,ether and ethanol,soluble in acetone | |
C(C(=O)O)(Cl)Cl | |
128.94 | |
CHEBI:36386 | |
Crystals or Crystalline Powder |
1.5600g/mL | |
Authentic | |
1.4653 to 1.4673 | |
194.0°C | |
9.0°C to 11.0°C | |
79-43-6 | |
C2H2Cl2O2 | |
MFCD00004223 | |
16,115 | |
acetic acid, dichloro, bichloracetic acid, bichloroacetic acid, dca acid, dichloracetic acid, dichlorethanoic acid, dichloro-acetic acid, dichloroacetic acid, dichloroethanoic acid, urner's liquid | |
JXTHNDFMNIQAHM-UHFFFAOYSA-N | |
2,2-dichloroacetic acid | |
6597 | |
128.94 | |
99+% |
Chemical Identifiers
79-43-6 | |
128.94 | |
JXTHNDFMNIQAHM-UHFFFAOYSA-N | |
6597 | |
2,2-dichloroacetic acid |
C2H2Cl2O2 | |
MFCD00004223 | |
acetic acid, dichloro, bichloracetic acid, bichloroacetic acid, dca acid, dichloracetic acid, dichlorethanoic acid, dichloro-acetic acid, dichloroacetic acid, dichloroethanoic acid, urner's liquid | |
CHEBI:36386 | |
C(C(=O)O)(Cl)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Suspected of causing cancer.
May damage fertility or the unborn child.
May cause harm to breast-fed children.
May cause damage to or
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 201-207-