Learn More
Dichloroacetic acid, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC113060050
Specifications
Dichloroacetic acid | |
Authentic | |
1.4650 to 1.4670 (20°C, 589nm) | |
5mL | |
99+% | |
Cl2CHCO2H | |
acetic acid, dichloro, bichloracetic acid, bichloroacetic acid, dca acid, dichloracetic acid, dichlorethanoic acid, dichloro-acetic acid, dichloroacetic acid, dichloroethanoic acid, urner's liquid | |
C(C(=O)O)(Cl)Cl | |
128.936 | |
CHEBI:36386 | |
Crystals or Crystalline Powder |
1.5600g/mL | |
Glass bottle | |
Brown | |
79-43-6 | |
C2H2Cl2O2 | |
MFCD00004223 | |
JXTHNDFMNIQAHM-UHFFFAOYSA-N | |
2,2-dichloroacetic acid | |
6597 | |
128.94 | |
≥99.0% (Acid) |
Chemical Identifiers
79-43-6 | |
128.936 | |
JXTHNDFMNIQAHM-UHFFFAOYSA-N | |
6597 | |
2,2-dichloroacetic acid |
C2H2Cl2O2 | |
MFCD00004223 | |
acetic acid, dichloro, bichloracetic acid, bichloroacetic acid, dca acid, dichloracetic acid, dichlorethanoic acid, dichloro-acetic acid, dichloroacetic acid, dichloroethanoic acid, urner's liquid | |
CHEBI:36386 | |
C(C(=O)O)(Cl)Cl |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger