missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloroacetic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113062500

 View more versions of this product

Catalog No. AC113062500



Description

Description

Specifications

Specifications

Dichloroacetic acid
>112°C
Glass bottle
1.56
Brown
250mL
99+%
Cl2CHCO2H
02,202
15,3059
Solubility in water: soluble. Other solubilities: miscible with alcohol,ether and ethanol,soluble in acetone
C(C(=O)O)(Cl)Cl
128.94
CHEBI:36386
Crystals or Crystalline Powder
1.5600g/mL
Authentic
1.4653 to 1.4673
194.0°C
9.0°C to 11.0°C
79-43-6
C2H2Cl2O2
MFCD00004223
16,115
acetic acid, dichloro, bichloracetic acid, bichloroacetic acid, dca acid, dichloracetic acid, dichlorethanoic acid, dichloro-acetic acid, dichloroacetic acid, dichloroethanoic acid, urner's liquid
JXTHNDFMNIQAHM-UHFFFAOYSA-N
2,2-dichloroacetic acid
6597
128.94
99+%

Chemical Identifiers

79-43-6
128.94
JXTHNDFMNIQAHM-UHFFFAOYSA-N
6597
2,2-dichloroacetic acid
C2H2Cl2O2
MFCD00004223
acetic acid, dichloro, bichloracetic acid, bichloroacetic acid, dca acid, dichloracetic acid, dichlorethanoic acid, dichloro-acetic acid, dichloroacetic acid, dichloroethanoic acid, urner's liquid
CHEBI:36386
C(C(=O)O)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Suspected of causing cancer.
May damage fertility or the unborn child.
May cause harm to breast-fed children.
May cause damage to or

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 201-207-

SDS Documents