Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloroacetic acid, 99%

Chemical Identifiers

Chemical Name or Material Dichloroacetic acid
CAS 79-43-6
Molecular Formula Cl2CHCO2H
Formula Weight 128.94
MDL Number MFCD00004223
UN Number UN1764
Beilstein 1098596
Merck Index 14,3050
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA147400E

  SDS
alfa aesar™
A147400E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1474022

  SDS
alfa aesar™
A1474022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1474036

  SDS
alfa aesar
A1474036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.94
Assay Percent Range 99%
Physical Form Liquid
Storage Note 1 Hygroscopic
UN Number UN1764
Assay 99%
Boiling Point 192° to 194°C
Melting Point 9° to 11°C
Quantity 2500g
Health Hazard 1 Danger
Flash Point >110°C (230°F)
TSCA TSCA
Merck Index 14,3050
CAS 79-43-6
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETIC ACID
RTECS Number AG6125000
Molecular Formula Cl2CHCO2H
Chemical Name or Material Dichloroacetic acid
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00004223
Beilstein 1098596
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Odor Pungent

Solubility
Miscible with water, ethanol, diethyl ether, n-octanol and acetone.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.

Hygroscopic