missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloroacetic acid, 99+%

$45.77 - $78.68

Chemical Identifiers

CAS 79-43-6
Formula Weight 128.94
MDL Number MFCD00004223
UN Number UN1764
Beilstein 1098596
Merck Index 14,3050
Chemical Name or Material Dichloroacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3124930 SDS Alfa Aesar™
3124930
250g Each for $45.77
AA31249A1 SDS Alfa Aesar™
31249A1
1kg Each for $78.68
Specifications Description & Specifications

Chemical Identifiers

79-43-6
MFCD00004223
1098596
Dichloroacetic acid
128.94
UN1764
14,3050

Specifications

>110° (230°F)
194°C
9° to 11°C
79-43-6
Pungent
Liquid
MFCD00004223
1098596
Dichloroacetic acid
Hygroscopic
Yellow
250g
1.564g/mL
1.4620
128.94
UN1764
14,3050
≥99%

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETIC ACID

RTECSNumber : AG6125000

TSCA : TSCA