missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloroacetic acid, 99+%

Manufacturer:  Alfa Aesar™ 31249A1

 View more versions of this product

Catalog No. AA31249A1



Specifications

Specifications

Dichloroacetic acid
>110° (230°F)
1.4620
194°C
9°C to 11°C
79-43-6
MFCD00004223
1098596
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
C(C(=O)O)(Cl)Cl
128.936
CHEBI:36386
Liquid
1.564g/mL
Pungent
Hygroscopic
Yellow
1kg
C2H2Cl2O2
UN1764
14,3050
JXTHNDFMNIQAHM-UHFFFAOYSA-N
2,2-dichloroacetic acid
6597
128.94
≥99%

Chemical Identifiers

79-43-6
128.936
JXTHNDFMNIQAHM-UHFFFAOYSA-N
6597
2,2-dichloroacetic acid
C2H2Cl2O2
MFCD00004223
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
CHEBI:36386
C(C(=O)O)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETIC ACID

RTECSNumber : AG6125000

TSCA : TSCA

SDS Documents
Promotions

Promotions