Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1-binaphthyl]palladium(II), 99%

$676.23 - $676.23

Chemical Identifiers

Chemical Name or Material Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1-binaphthyl]palladium(II)
CAS 115826-95-4
Molecular Formula C44H32Cl2P2Pd
Formula Weight 800.01g/mol
MDL Number MFCD00075254
Synonym [(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II) chloride; PdCl2((R)-(+)-BINAP)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2780903

 
alfa aesar™
H27809-03
1g Each for $676.23
Description & Specifications

Specifications

Quantity 1g
Optical Rotation +676° (c=0.5 in Chloroform)
Chemical Name or Material Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1-binaphthyl]palladium(II)
CAS 115826-95-4
Assay 99%
Molecular Formula C44H32Cl2P2Pd
Formula Weight 800.01g/mol
MDL Number MFCD00075254
Synonym [(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II) chloride; PdCl2((R)-(+)-BINAP)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.