Please login to your online account to display your discounted pricing

Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
CAS 96503-27-4
Molecular Formula (C10H15Cl2Ru)n
MDL Number MFCD00070449
Synonym Pentamethylcyclopentadienylruthenium(III) chloride polymer
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363322500

  SDS
thermo fisher scientific
363322500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 250mg
Color Red to very dark brown
Synonym Pentamethylcyclopentadienylruthenium(III) chloride polymer
Packaging Glass bottle
Infrared Spectrum Authentic
Elemental Analysis 97.5% min.
Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
MDL Number MFCD00070449
Molecular Formula (C10H15Cl2Ru)n
CAS 96503-27-4