Please login to your online account to display your discounted pricing

Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  363322500

 View more versions of this product

Catalog No. AC363322500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 250mg
Color Red to very dark brown
Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Molecular Formula (C10H15Cl2Ru)n
MDL Number MFCD00070449
Packaging Glass bottle
CAS 96503-27-4
Synonym Pentamethylcyclopentadienylruthenium(III) chloride polymer

Specifications

Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
CAS 96503-27-4
Molecular Formula (C10H15Cl2Ru)n
MDL Number MFCD00070449
Synonym Pentamethylcyclopentadienylruthenium(III) chloride polymer