Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer

Chemical Identifiers

Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
CAS 96503-27-4
Formula Weight 307.21
MDL Number MFCD00070449
Synonym (RuCl2Cp*)n
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4634803

  SDS
alfa aesar™
4634803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
Sensitivity Air & Moisture Sensitive
Color Red-brown
Melting Point 45° to 47°C
Quantity 1g
CAS 96503-27-4
Storage Note 1 2° to 8°C
Physical Form Crystalline Solid
Formula Weight 307.21
MDL Number MFCD00070449
Synonym (RuCl2Cp*)n
Safety and Handling

2° to 8°C