Please login to your online account to display your discounted pricing

α,α'-Dichloro-o-xylene 98%, ACROS Organics™

$134.59 - $134.59

Chemical Identifiers

Chemical Name or Material α, α'-Dichloro-o-xylene
Name Note 98%
CAS 612-12-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8Cl2
Linear Formula C6H4(CH2Cl)2
Formula Weight 175.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC154141000

  SDS
thermo fisher scientific
154141000
Glass bottle 100g Each for $134.59
Description & Specifications

Specifications

Physical Form Crystals or Crystalline Powder
Assay ≥97.5%
Formula Weight 175.06
Quantity 100g
Color White semi-transparent to light beige
Beilstein 05, 364
EINECS Number 210-291-8
TSCA TSCA
Melting Point 53° to 57°C
RTECS Number ZE4085000
CAS Max % 100.0
Chemical Name or Material α, α'-Dichloro-o-xylene
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H8Cl2
Linear Formula C6H4(CH2Cl)2
MDL Number MFCD00000903
CAS 612-12-4
Synonym 1, 2-Bis(chloromethyl)benzene
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.