missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha'-Dichloro-o-xylene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 154141000

Catalog No. AC154141000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

α, α'-Dichloro-o-xylene
612-12-4
100.0
Beige to White
107°C
97.5% min. (GC)
C8H8Cl2
MFCD00000903
05, 364
Solubility in water: hydrolysis
ClCC1=CC=CC=C1CCl
175.05
175.06
Crystalline Powder or Crystals
98%
97.5
53°C to 57°C
239°C to 241°C
Authentic
Glass bottle
C6H4(CH2Cl)2
100 g
1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid
FMGGHNGKHRCJLL-UHFFFAOYSA-N
1,2-bis(chloromethyl)benzene
11919
98%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 210-291-8

RTECSNumber : ZE4085000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only