missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro(ethylenediamine)platinum(II), Pt 59.7% min

$125.51 - $384.12

Chemical Identifiers

CAS 14096-51-6
MDL Number MFCD00000011
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1049701
View Documents
Alfa Aesar™
1049701
0.25g
Each for $125.51
 
AA1049703
View Documents
Alfa Aesar™
1049703
1g
Each for $384.12
 
Description

Description

Specifications

Chemical Identifiers

14096-51-6
MFCD00000011

Specifications

Hygroscopic
14096-51-6
Pt 59.7% min.
326.1
Dichloro(ethylenediamine)platinum(II)
0.25g
MFCD00000011
Soluble in water.
Crystalline/Powder
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-943-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS