missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro(ethylenediamine)platinum(II), Pt 59.7% min

Manufacturer:  Alfa Aesar™ 1049703

 View more versions of this product

Catalog No. AA1049703



Specifications

Specifications

Dichloro(ethylenediamine)platinum(II)
1g
MFCD00000011
Soluble in water.
Crystalline/Powder
Hygroscopic
14096-51-6
Pt 59.7% min.
326.1

Chemical Identifiers

14096-51-6
MFCD00000011
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-943-4

TSCA : No

Recommended Storage : Ambient temperatures

Documents