missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro(ethylenediamine)palladium(II), Pd 44.8%

Manufacturer:  Alfa Aesar™ 1049606

 View more versions of this product

Catalog No. AA1049606



Specifications

Specifications

Dichloro(ethylenediamine)palladium(II)
5g
C2H8Cl2N2Pd
dichloro 1,2-diaminoethane ;palladium
CAYKJANQVKIYPJ-UHFFFAOYSA-L
dichloropalladium;ethane-1,2-diamine
11139157
Crystalline
Hygroscopic
15020-99-2
MFCD00044991
Pd 44.8%
C(CN)N.Cl[Pd]Cl
237.42
237.41

Chemical Identifiers

15020-99-2
237.42
CAYKJANQVKIYPJ-UHFFFAOYSA-L
11139157
C(CN)N.Cl[Pd]Cl
C2H8Cl2N2Pd
MFCD00044991
dichloro 1,2-diaminoethane ;palladium
dichloropalladium;ethane-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents