Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro(dimethylamino)phosphine, 98%

$197.03 - $197.03

Chemical Identifiers

Chemical Name or Material Dichloro(dimethylamino)phosphine
CAS 683-85-2
Molecular Formula C2H6Cl2NP
Linear Formula (CH3)2NPCl2
Formula Weight 145.96
MDL Number MFCD00013613
UN Number UN2924
Synonym (Dimethylamino)dichlorophosphine; Dimethylphosphoramidous dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2785606

 
alfa aesar™
H2785606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2785614

 
alfa aesar™
H2785614
25g Each for $197.03
Description & Specifications

Specifications

Formula Weight 145.96
Chemical Name or Material Dichloro(dimethylamino)phosphine
Assay 98%
Molecular Formula C2H6Cl2NP
Linear Formula (CH3)2NPCl2
MDL Number MFCD00013613
CAS 683-85-2
UN Number UN2924
Boiling Point 150°C
Flash Point −23°C (−9°F)
Synonym (Dimethylamino)dichlorophosphine; Dimethylphosphoramidous dichloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Quantity 5g

Applications
Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following N,N-dimethylamidotetrachlorophosphorane, O-propylchloroformimino-N,N-dimethylamidochlorophosphate, bisarylsulphonyldiazadiphosphetidines.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C