Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[bis(dicyclohexylphosphinophenyl)ether]palladium(II), 98% min

Chemical Identifiers

Chemical Name or Material Dichloro[bis(dicyclohexylphosphinophenyl)ether]palladium(II)
CAS 1265140-81-5
Formula Weight 740.08
MDL Number MFCD26967909
Synonym [Bis(dicyclohexylphosphinophenyl)ether]dichloropalladium(II); [Bis(dicyclohexylphosphinophenyl)ether]palladium(II) dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4687203

  SDS
alfa aesar™
4687203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 740.08
Chemical Name or Material Dichloro[bis(dicyclohexylphosphinophenyl)ether]palladium(II)
Assay ≥98%
Color Yellow
Physical Form Solid
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD26967909
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1265140-81-5
Synonym [Bis(dicyclohexylphosphinophenyl)ether]dichloropalladium(II); [Bis(dicyclohexylphosphinophenyl)ether]palladium(II) dichloride
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.