Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[bis(1,3-diphenylphosphino)propane]palladium(II)

$135.61 - $339.03

Chemical Identifiers

Chemical Name or Material Dichloro[bis(1,3-diphenylphosphino)propane]palladium(II)
CAS 59831-02-6
Molecular Formula C27H26Cl2P2Pd
Formula Weight 589.77
MDL Number MFCD03844773
Synonym [1,3-Bis(diphenylphosphino)propane]dichloropalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2689703

  SDS
alfa aesar™
H2689703
1g Each for $339.03

AAH26897MD

  SDS
alfa aesar™
H26897MD
250mg Each for $135.61
Description & Specifications

Specifications

Formula Weight 589.77
Chemical Name or Material Dichloro[bis(1,3-diphenylphosphino)propane]palladium(II)
Molecular Formula C27H26Cl2P2Pd
MDL Number MFCD03844773
CAS 59831-02-6
Synonym [1,3-Bis(diphenylphosphino)propane]dichloropalladium(II)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
(1,3-Bis(diphenylphosphino)propane)palladium(II) chloride, used ubiquitously as a employed preparation of palladium for cross-coupling protocols in organic synthesis. It is also a useful organo-metallic chemical catalyst. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in coupling reactions.

Solubility
Soluble in dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.