missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), Alfa Aesar™

Manufacturer:  Alfa Aesar 3016709

 View more versions of this product

Catalog No. AA3016709



Specifications

Specifications

Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II)
10g
C27H28Cl2NiP2
1,3-Bis(diphenylphosphino)propane nickel(II) chloride
C1=CC=C(C=C1)P(CCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni]
544.062
542.03
213°C (decomposition)
15629-92-2
MFCD00015318
BILYTLYIOVUVSU-UHFFFAOYSA-N
3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
131675641
Powder

Chemical Identifiers

15629-92-2
544.062
BILYTLYIOVUVSU-UHFFFAOYSA-N
131675641
C1=CC=C(C=C1)P(CCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni]
C27H28Cl2NiP2
MFCD00015318
1,3-Bis(diphenylphosphino)propane nickel(II) chloride
3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H317-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents