Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II), 98%

$56.50 - $170.27

Chemical Identifiers

Chemical Name or Material Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)
CAS 14647-23-5
Formula Weight 528.02
MDL Number MFCD00013313
Synonym 1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1393204

  SDS
alfa aesar™
1393204
2g Each for $56.50

AA1393209

  SDS
alfa aesar™
1393209
10g Each for $170.27
Description & Specifications

Specifications

Formula Weight 528.02
Health Hazard 2 GHS H Statement
H351-H302-H312-H332-H315-H319-H317-H335
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Synonym 1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
Color Orange-red to brown
Melting Point 263° to 265°C
Chemical Name or Material Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)
Physical Form Powder
Assay 98%
CAS 14647-23-5
Quantity 2g
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
MDL Number MFCD00013313
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H302-H312-H332-H315-H319-H317-H335
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.