missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II), 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1393209

 View more versions of this product

Catalog No. AA1393209



Specifications

Specifications

Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)
263°C to 265°C
14647-23-5
MFCD00013313
XXECWTBMGGXMKP-UHFFFAOYSA-N
131664338
Powder
Brown to Orange-Red
10g
C26H26Cl2NiP2+2
1,2-bis diphenylphosphino ethane nickel ii chloride
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni+2]
528.02
98%

Chemical Identifiers

14647-23-5
MFCD00013313
1,2-bis diphenylphosphino ethane nickel ii chloride
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni+2]
C26H26Cl2NiP2+2
XXECWTBMGGXMKP-UHFFFAOYSA-N
131664338
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302-H312-H332-H315-H319-H317-H335
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents