missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II), 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5502014

 View more versions of this product

Catalog No. AAH5502014



Description

Description

Applications
Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II) is used as a catalyst for regio- and stereoselective intermolecular enzyme coupling, cross-coupling reactions of alkyl halides with aryl Grignard reagents, hydrovinylation of styrene, tandem radical cyclization and cross-coupling, dimethylation, hydrodechlorination and addition reactions.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas. Protect from humidity and water.
Specifications

Specifications

Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II)
260°C (decomposition)
18498-01-6
CoCl2[(C6H5)2PCH2CH2P(C6H5)2]
1,2-bis diphenylphosphino ethane cobaltdichloride
SYTWXWRJCLAZFP-UHFFFAOYSA-L
dichlorocobalt;2-diphenylphosphanylethyl(diphenyl)phosphane
11005917
97%
Hygroscopic
25g
C26H24Cl2CoP2
MFCD00134173
Insoluble in water.
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[Co]Cl
528.259
528.26

Chemical Identifiers

18498-01-6
528.259
SYTWXWRJCLAZFP-UHFFFAOYSA-L
11005917
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[Co]Cl
C26H24Cl2CoP2
MFCD00134173
1,2-bis diphenylphosphino ethane cobaltdichloride
dichlorocobalt;2-diphenylphosphanylethyl(diphenyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

Documents