missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 14.1%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4666503

 View more versions of this product

Catalog No. AA4666503



Specifications

Specifications

Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)
280°C to 287°C (decomposition)
205319-10-4
MFCD14155707
Pd 14.1%
CC1(C2=C(C(=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)OC5=C1C=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7)C.[Cl-].[Cl-].[Pd+2]
755.952
755.96
Yellow
1g
C39H32Cl2OP2Pd
dichloro 9,9-dimethyl-4,5-bis diphenylphosphino xanthene palladium ii, palladium 2+ xantphos dichloride, 4,5-bis diphenylphosphino-9,9-dimethylxanthene dichloropalladium ii, 4,5-bis diphenylphosphino-9,9-dimethylxanthene palladium ii dichloride
HEYONDYPXIUDCK-UHFFFAOYSA-L
(5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane;palladium(2+);dichloride
86280406
Crystalline Solid

Chemical Identifiers

205319-10-4
755.952
HEYONDYPXIUDCK-UHFFFAOYSA-L
86280406
CC1(C2=C(C(=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)OC5=C1C=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7)C.[Cl-].[Cl-].[Pd+2]
C39H32Cl2OP2Pd
MFCD14155707
dichloro 9,9-dimethyl-4,5-bis diphenylphosphino xanthene palladium ii, palladium 2+ xantphos dichloride, 4,5-bis diphenylphosphino-9,9-dimethylxanthene dichloropalladium ii, 4,5-bis diphenylphosphino-9,9-dimethylxanthene palladium ii dichloride
(5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane;palladium(2+);dichloride
SDS Documents